accessibility menu, dialog, popup

UserName

Viewing profile as user:
PPPPNi

CAS RN 15133-82-1

IUPAC Name: tetrakis(triphenylphosphane) nickel
Synonyms: tetrakis(triphenylphosphine) nickel
Molecular Weight (g/mol): 1107.86
Molecular Formula: C72H60NiP4
InChi Key: KFBKRCXOTTUAFS-UHFFFAOYSA-N
SMILES: [Ni].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)