accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15139-39-6

IUPAC Name: 2,4-dimethylfuran-3-carbonyl chloride
Synonyms: 2,4-dimethylfuran-3-carbonyl chloride
Molecular Weight (g/mol): 158.58
Molecular Formula: C7H7ClO2
InChi Key: NOEQXZGBSMGWCT-UHFFFAOYSA-N
SMILES: CC1=C(C(Cl)=O)C(C)=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results