accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15145-69-4

IUPAC Name: 1-chloro-3-(dichloromethyl)benzene
Synonyms: 1-chloro-3-(dichloromethyl)benzene
Molecular Weight (g/mol): 195.47
Molecular Formula: C7H5Cl3
InChi Key: ACNQJGLSENYFQJ-UHFFFAOYSA-N
SMILES: ClC(Cl)C1=CC=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results