accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH2(E)

CAS RN 1515-76-0

IUPAC Name: (1E)-buta-1,3-dien-1-yl acetate
Synonyms: (1E)-buta-1,3-dien-1-yl acetate
Molecular Weight (g/mol): 112.13
Molecular Formula: C6H8O2
InChi Key: NMQQBXHZBNUXGJ-SNAWJCMRSA-N
SMILES: CC(=O)O\C=C\C=C
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results