accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1515-95-3

IUPAC Name: 1-ethyl-4-methoxybenzene
Synonyms: benzene, 1-ethyl-4-methoxy-
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: HDNRAPAFJLXKBV-UHFFFAOYSA-N
SMILES: CCC1=CC=C(OC)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)