accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15159-40-7

IUPAC Name: morpholine-4-carbonyl chloride
Synonyms: morpholine-4-carbonyl chloride
Molecular Weight (g/mol): 149.57
Molecular Formula: C5H8ClNO2
InChi Key: XMWFMEYDRNJSOO-UHFFFAOYSA-N
SMILES: ClC(=O)N1CCOCC1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)