accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15163-30-1

IUPAC Name: 4-(dodecyldimethylazaniumyl)butanoate
Synonyms: 4-(dodecyldimethylammonio)butanoate
Molecular Weight (g/mol): 299.5
Molecular Formula: C18H37NO2
InChi Key: ZFVNBVQBIZJUEY-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC[N+](C)(C)CCCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results