accessibility menu, dialog, popup

UserName

Viewing profile as user:
IO

CAS RN 15164-44-0

IUPAC Name: 4-iodobenzaldehyde
Synonyms: 4-iodobenzaldehyde
Molecular Weight (g/mol): 232.02
Molecular Formula: C7H5IO
InChi Key: NIEBHDXUIJSHSL-UHFFFAOYSA-N
SMILES: IC1=CC=C(C=O)C=C1
Percent Purity 
  • (2)
  • (3)
  • (1)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)