accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15174-69-3

IUPAC Name: 4-hydroxy-3-methylbenzaldehyde
Synonyms: 4-hydroxy-3-methylbenzaldehyde
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: BAKYASSDAXQKKY-UHFFFAOYSA-N
SMILES: CC1=CC(C=O)=CC=C1O
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results