accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1518-72-5

IUPAC Name: 2-methyl-1-[(2-methylpropyl)disulfanyl]propane
Synonyms: 2-methyl-1-[(2-methylpropyl)disulfanyl]propane
Molecular Weight (g/mol): 178.35
Molecular Formula: C8H18S2
InChi Key: ONJROLGQWMBXAP-UHFFFAOYSA-N
SMILES: CC(C)CSSCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results