accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1518-84-9

IUPAC Name: 2-cyclopentylphenol
Synonyms: 2-cyclopentylphenol
Molecular Weight (g/mol): 162.23
Molecular Formula: C11H14O
InChi Key: JHEKSKQMOBLXQS-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C1CCCC1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)