accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15182-06-6

IUPAC Name: 2-[2-(dimethylamino)ethoxy]benzaldehyde
Synonyms: 2-[2-(dimethylamino)ethoxy]benzaldehyde
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: TWMYSXRSVLFCGX-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results