accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSSOOCl

CAS RN 151858-64-9

IUPAC Name: 5-(pyridin-2-yl)thiophene-2-sulfonyl chloride
Synonyms: 5-(pyridin-2-yl)thiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 259.72
Molecular Formula: C9H6ClNO2S2
InChi Key: OGOMWPWAEIDEOU-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(S1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results