CAS RN 15187-16-3
IUPAC Name:
λ²-lead(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide
Synonyms:
λ²-lead(2+) 2,11,20,29,37,38,39,40-octaazanonacyclo[28.6.1.1³,¹⁰.1¹²,¹⁹.1²¹,²⁸.0⁴,⁹.0¹³,¹⁸.0²²,²⁷.0³¹,³⁶]tetraconta-1(36),2,4,6,8,10(40),11,13,15,17,19(39),20,22,24,26,28,30,32,34-nonadecaene-37,38-diide
Molecular Weight (g/mol):
719.7
Molecular Formula:
C32H16N8Pb
InChi Key:
WSQYJDCCFQPFJC-UHFFFAOYSA-N
SMILES:
[Pb++].[N-]1C2=NC3=C4C=CC=CC4=C([N-]3)N=C3N=C(N=C4N=C(N=C1C1=CC=CC=C21)C1=CC=CC=C41)C1=CC=CC=C31
1
–
2
of
2
results
1
Lead(II) Phthalocyanine 95.0+%, TCI America™
CAS: 15187-16-3 Molecular Formula: C32H16N8Pb MDL Number: MFCD00053951 Synonym: Phthalocyanine Lead(II)
2
Lead(II) Phthalocyanine (purified by sublimation) 98.0+%, TCI America™
CAS: 15187-16-3 Molecular Formula: C32H16N8Pb MDL Number: MFCD00053951 Synonym: Phthalocyanine Lead(II)