accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1519-45-5

IUPAC Name: triphenyl[2-(triphenylphosphaniumyl)ethyl]phosphanium diiodide
Synonyms: triphenyl[2-(triphenylphosphaniumyl)ethyl]phosphanium diiodide
Molecular Weight (g/mol): 806.45
Molecular Formula: C38H34I2P2
InChi Key: VKWZUCXGFNDLSS-UHFFFAOYSA-L
SMILES: [I-].[I-].C(C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results