accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHONHNHNO

CAS RN 15191-21-6

IUPAC Name: 3-(1H-imidazol-5-yl)-2-formamidopropanoic acid
Synonyms: 3-(3H-imidazol-4-yl)-2-formamidopropanoic acid
Molecular Weight (g/mol): 183.17
Molecular Formula: C7H9N3O3
InChi Key: WFQVSLVQIFFQQN-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CN=CN1)NC=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results