accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15199-41-4

IUPAC Name: cyclododecane-1,2-diol
Synonyms: cyclododecane-1,2-diol
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: HAMFVYJFVXTJCJ-UHFFFAOYNA-N
SMILES: OC1CCCCCCCCCCC1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results