accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSOOO2H2H2H2H2H2H

CAS RN 15199-43-6

IUPAC Name: di(²H₃)methyl sulfate
Synonyms: di(²H₃)methyl sulfate
Molecular Weight (g/mol): 132.16
Molecular Formula: C2H6O4S
InChi Key: VAYGXNSJCAHWJZ-WFGJKAKNSA-N
SMILES: [2H]C([2H])([2H])OS(=O)(=O)OC([2H])([2H])[2H]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results