accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)(S)(R)O(S)(R)HOHC

CAS RN 152-43-2

IUPAC Name: (1R,3aS,3bR,9bS,11aS)-7-(cyclopentyloxy)-1-ethynyl-11a-methyl-1H,2H,3H,3aH,3bH,4H,5H,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-ol
Synonyms: quinestrol quinestrol (product) plestrovis estrovis ethinyl estradiol 3 cyclopentyl ether qui-lea quinestrolo eston quilea estrovister
Molecular Weight (g/mol): 364.53
Molecular Formula: C25H32O2
InChi Key: PWZUUYSISTUNDW-VAFBSOEGSA-N
SMILES: C[C@]12CC[C@H]3[C@@H](CCC4=CC(OC5CCCC5)=CC=C34)[C@@H]1CC[C@@]2(O)C#C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results