accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOHOHOOOHOHOHOH

CAS RN 152-95-4

IUPAC Name: 5,7-dihydroxy-3-(4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phenyl)-4H-chromen-4-one
Synonyms: sophoricoside
Molecular Weight (g/mol): 432.38
Molecular Formula: C21H20O10
InChi Key: ISQRJFLLIDGZEP-UHFFFAOYNA-N
SMILES: OCC1OC(OC2=CC=C(C=C2)C2=COC3=CC(O)=CC(O)=C3C2=O)C(O)C(O)C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results