accessibility menu, dialog, popup

UserName

Viewing profile as user:
2H2H2H2H2H2H2H2H2H2H2H2H

CAS RN 1520-96-3

IUPAC Name: (²H₁₂)perylene
Synonyms: (²H₁₂)perylene
Molecular Weight (g/mol): 264.39
Molecular Formula: C20H12
InChi Key: CSHWQDPOILHKBI-AQZSQYOVSA-N
SMILES: [2H]C1=C([2H])C2=C3C(C([2H])=C([2H])C([2H])=C3C3=C4C(C([2H])=C([2H])C([2H])=C24)=C([2H])C([2H])=C3[2H])=C1[2H]
Solvent or Matrix 
  • (1)
  • (1)
Element 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)