accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 15206-55-0

IUPAC Name: methyl 2-oxo-2-phenylacetate
Synonyms: methyl benzoylformate
Molecular Weight (g/mol): 164.16
Molecular Formula: C9H8O3
InChi Key: YLHXLHGIAMFFBU-UHFFFAOYSA-N
SMILES: COC(=O)C(=O)C1=CC=CC=C1
Boiling Point 
  • (3)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (7)