accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNH2HNN

CAS RN 1520864-90-7

IUPAC Name: 1H-1,3-benzodiazole-6-carbothioamide
Synonyms: 3H-1,3-benzodiazole-5-carbothioamide
Molecular Weight (g/mol): 177.23
Molecular Formula: C8H7N3S
InChi Key: SWADWMCPVIOXDO-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C2N=CNC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results