accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1521-51-3

IUPAC Name: 3-bromocyclohex-1-ene
Synonyms: 3-bromocyclohex-1-ene
Molecular Weight (g/mol): 161.04
Molecular Formula: C6H9Br
InChi Key: AJKDUJRRWLQXHM-UHFFFAOYNA-N
SMILES: BrC1CCCC=C1
Boiling Point 
  • (1)
  • (3)
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (2)
  • (6)
  • (1)
  • (3)
  • (3)
  • (2)
  • (4)
  • (5)
  • (2)
  • (1)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)