accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1522-88-9

IUPAC Name: 1,3-diiodo-2,2-bis(iodomethyl)propane
Synonyms: 1,3-diiodo-2,2-bis(iodomethyl)propane
Molecular Weight (g/mol): 575.74
Molecular Formula: C5H8I4
InChi Key: HPMGSGCDKVCZHC-UHFFFAOYSA-N
SMILES: ICC(CI)(CI)CI
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results