accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15230-79-2

IUPAC Name: lutetium(3+) tris(nitric acid)
Synonyms: lutetium(3+) tris(nitric acid)
Molecular Weight (g/mol): 364
Molecular Formula: H3LuN3O9
InChi Key: MESQZVHAWZZUAY-UHFFFAOYSA-N
SMILES: [Lu+3].O[N+]([O-])=O.O[N+]([O-])=O.O[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)