accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOOO

CAS RN 15231-78-4

IUPAC Name: 2,2-dimethyl-5-phenyl-1,3-dioxane-4,6-dione
Synonyms: 2,2-dimethyl-5-phenyl-1,3-dioxane-4,6-dione
Molecular Weight (g/mol): 220.22
Molecular Formula: C12H12O4
InChi Key: NTUAHNQWHPQAMB-UHFFFAOYSA-N
SMILES: CC1(C)OC(=O)C(C(=O)O1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)