accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15233-47-3

IUPAC Name: N1-(octan-2-yl)-N4-phenylbenzene-1,4-diamine
Synonyms: N1-(octan-2-yl)-N4-phenylbenzene-1,4-diamine
Molecular Weight (g/mol): 296.46
Molecular Formula: C20H28N2
InChi Key: JQTYAZKTBXWQOM-UHFFFAOYNA-N
SMILES: CCCCCCC(C)NC1=CC=C(NC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results