accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15244-35-6

IUPAC Name: cadmium(2+) sulfate
Synonyms: cadmium sulfate|CdSO4|cadmiumsulfat|sulfate de cadmium|sulfato de cadmio|kadmiumsulfat
Molecular Weight (g/mol): 208.47
Molecular Formula: CdO4S
InChi Key: QCUOBSQYDGUHHT-UHFFFAOYSA-L
SMILES: [Cd++].[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)