accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15244-36-7

IUPAC Name: manganese(2+) hydrate sulfate
Synonyms: manganese(2+) hydrate sulfate
Molecular Weight (g/mol): 169.01
Molecular Formula: H2MnO5S
InChi Key: ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES: O.[Mn++].[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)