accessibility menu, dialog, popup

UserName

Viewing profile as user:
CrOSOOOOSOOOOSOOOCr

CAS RN 15244-38-9

IUPAC Name: dichromium(3+) trisulfate
Synonyms: chromium sulphate|chromium sulfate|chromic sulfate|chromium(III) sulfate|chromic sulphate|dichromium sulfate|dichromium trisulfate|dichromium sulphate|chronisulfat|dichromium trisulphate
Molecular Weight (g/mol): 392.16
Molecular Formula: Cr2O12S3
InChi Key: GRWVQDDAKZFPFI-UHFFFAOYSA-H
SMILES: [Cr+3].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)