accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15260-10-3

IUPAC Name: (2S,3R)-3-(benzyloxy)-2-{[(tert-butoxy)carbonyl]amino}butanoic acid
Synonyms: (2S,3R)-3-(benzyloxy)-2-[(tert-butoxycarbonyl)amino]butanoic acid|BOC-THR(BZL)-OH
Molecular Weight (g/mol): 309.36
Molecular Formula: C16H23NO5
InChi Key: CTXPLTPDOISPTE-YPMHNXCESA-N
SMILES: C[C@@H](OCC1=CC=CC=C1)[C@H](NC(=O)OC(C)(C)C)C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)