accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSiClOH3COH3C

CAS RN 15267-95-5

IUPAC Name: (chloromethyl)triethoxysilane
Synonyms: (chloromethyl)triethoxysilane
Molecular Weight (g/mol): 212.75
Molecular Formula: C7H17ClO3Si
InChi Key: ZDOBWJOCPDIBRZ-UHFFFAOYSA-N
SMILES: CCO[Si](CCl)(OCC)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results