accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 152684-30-5

IUPAC Name: 5-bromo-2-methoxy-3-nitropyridine
Synonyms: 5-bromo-2-methoxy-3-nitropyridine
Molecular Weight (g/mol): 233.02
Molecular Formula: C6H5BrN2O3
InChi Key: YRVHFGOAEVWBNS-UHFFFAOYSA-N
SMILES: COC1=NC=C(Br)C=C1[N+]([O-])=O
Quantity 
  • (1)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)