accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1527-89-5

IUPAC Name: 3-methoxybenzonitrile
Synonyms: 3-methoxybenzonitrile
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: KLXSUMLEPNAZFK-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1)C#N
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)