accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1527-91-9

IUPAC Name: [phenyl(phenylamino)methylidene]azanium
Synonyms: [phenyl(phenylamino)methylidene]azanium
Molecular Weight (g/mol): 197.26
Molecular Formula: C13H13N2
InChi Key: MPYOKHFSBKUKPQ-UHFFFAOYSA-O
SMILES: [NH2+]=C(NC1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results