accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1527-96-4

IUPAC Name: 1-(but-2-en-1-ylidene)-2-(2,4-dinitrophenyl)hydrazine
Synonyms: 1-(but-2-en-1-ylidene)-2-(2,4-dinitrophenyl)hydrazine
Molecular Weight (g/mol): 250.21
Molecular Formula: C10H10N4O4
InChi Key: GFUVNGJSSAEZHW-UHFFFAOYSA-N
SMILES: CC=CC=NNC1=C(C=C(C=C1)[N+]([O-])=O)[N+]([O-])=O
Concentration 
  • (1)
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)