accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH2

CAS RN 1528-30-9

IUPAC Name: methylidenecyclopentane
Synonyms: cyclopentane, methylene-
Molecular Weight (g/mol): 82.15
Molecular Formula: C6H10
InChi Key: NFJPEKRRHIYYES-UHFFFAOYSA-N
SMILES: C=C1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results