accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15280-31-6

IUPAC Name: zinc(2+) bis(4-[benzyl(ethyl)amino]benzene-1-diazonium) tetrachloride
Synonyms: zinc(2+) bis(4-[benzyl(ethyl)amino]benzenediazonium) tetrachloride
Molecular Weight (g/mol): 683.81
Molecular Formula: C30H32Cl4N6Zn
InChi Key: GZWAXOPGCCVFJG-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Zn++].CCN(CC1=CC=CC=C1)C1=CC=C(C=C1)[N+]#N.CCN(CC1=CC=CC=C1)C1=CC=C(C=C1)[N+]#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results