accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15280-55-4

CH3OOCH3OOCH3OODyH2OH2OH2OH2O

CAS RN 15280-55-4

IUPAC Name: dysprosium(3+) triacetate tetrahydrate
Synonyms: dysprosium(3+) triacetate tetrahydrate
Molecular Weight (g/mol): 411.69
Molecular Formula: C6H17DyO10
InChi Key: SIYZPSYZNCQFDV-UHFFFAOYSA-K
SMILES: O.O.O.O.[Dy+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Percent Purity 
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (4)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)