CAS RN 152803-24-2
IUPAC Name:
5-hydroxypyridine-3-carbonitrile
Synonyms:
5-hydroxypyridine-3-carbonitrile
Molecular Weight (g/mol):
120.11
Molecular Formula:
C6H4N2O
InChi Key:
KAXIYYPIORYZLB-UHFFFAOYSA-N
SMILES:
OC1=CN=CC(=C1)C#N
1
–
1
of
1
results
1
Promotions Available
3-Cyano-5-hydroxypyridine, 97%
3-Cyano-5-hydroxypyridine, 97%
CAS: 152803-24-2 Molecular Formula: C6H4N2O Molecular Weight (g/mol): 120.11 MDL Number: MFCD11044307 InChI Key: KAXIYYPIORYZLB-UHFFFAOYSA-N PubChem CID: 15342142 IUPAC Name: 5-hydroxypyridine-3-carbonitrile SMILES: OC1=CN=CC(=C1)C#N