accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONCl

CAS RN 152820-13-8

IUPAC Name: ethyl 4-chloropiperidine-1-carboxylate
Synonyms: ethyl 4-chloropiperidine-1-carboxylate
Molecular Weight (g/mol): 191.66
Molecular Formula: C8H14ClNO2
InChi Key: VRTIIQLEEGGJNE-UHFFFAOYSA-N
SMILES: CCOC(=O)N1CCC(Cl)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results