accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15283-45-1

IUPAC Name: {[(auriooxy)sulfonyl]sulfanyl}sodium; {[(sodiooxy)sulfonyl]sulfanyl}sodium
Synonyms: [(auriooxysulfonyl)sulfanyl]sodium; [(sodiooxysulfonyl)sulfanyl]sodium
Molecular Weight (g/mol): 490.17
Molecular Formula: AuNa3O6S4
InChi Key: FJCMCHNDCIYVST-UHFFFAOYSA-J
SMILES: [Na]OS(=O)(=O)S[Na].[Na]SS(=O)(=O)O[Au]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results