accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOOOOOOOEuEu

CAS RN 152864-32-9

IUPAC Name: dieuropium(3+) trioxalate
Synonyms: dieuropium(3+) trioxalate
Molecular Weight (g/mol): 567.98
Molecular Formula: C6Eu2O12
InChi Key: PVDYMOCCGHXJAK-UHFFFAOYSA-H
SMILES: [Eu+3].[Eu+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results