accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1528757-88-1

IUPAC Name: 2-[3-(benzyloxy)-5-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-[3-(benzyloxy)-5-fluorophenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 328.19
Molecular Formula: C19H22BFO3
InChi Key: FAVUCVNUXHFOPD-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(OCC2=CC=CC=C2)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results