accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1529-41-5

IUPAC Name: 2-(3-chlorophenyl)acetonitrile
Synonyms: 2-(3-chlorophenyl)acetonitrile
Molecular Weight (g/mol): 151.59
Molecular Formula: C8H6ClN
InChi Key: GTIKLPYCSAMPNG-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(CC#N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results