CAS RN 15290-77-4
IUPAC Name:
1,1,2,2,3,3,4-heptafluorocyclopentane
Synonyms:
1,1,2,2,3,3,4-heptafluorocyclopentane
Molecular Weight (g/mol):
196.07
Molecular Formula:
C5H3F7
InChi Key:
IDBYQQQHBYGLEQ-UHFFFAOYNA-N
SMILES:
FC1CC(F)(F)C(F)(F)C1(F)F
1
–
1
of
1
results
1
1,1,2,2,3,3,4-Heptafluorocyclopentane 98.0+%, TCI America™
CAS: 15290-77-4 Molecular Formula: C5H3F7 Molecular Weight (g/mol): 196.068 MDL Number: MFCD03844796 InChI Key: IDBYQQQHBYGLEQ-UHFFFAOYSA-N PubChem CID: 3853245 IUPAC Name: 1,1,2,2,3,3,4-heptafluorocyclopentane SMILES: C1C(C(C(C1(F)F)(F)F)(F)F)F