accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOHHO

CAS RN 152915-83-8

IUPAC Name: [6-(benzyloxy)naphthalen-2-yl]boronic acid
Synonyms: 6-(benzyloxy)naphthalen-2-ylboronic acid
Molecular Weight (g/mol): 278.11
Molecular Formula: C17H15BO3
InChi Key: ZVHAZMUBLCEMGG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2C=C(OCC3=CC=CC=C3)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results