accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 15294-63-0

IUPAC Name: tributyl(dodecyl)phosphanium bromide
Synonyms: tributyl(dodecyl)phosphanium bromide
Molecular Weight (g/mol): 451.56
Molecular Formula: C24H52BrP
InChi Key: ABJGUZZWSKMTEI-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCC[P+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results