accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OH3CO

CAS RN 15299-99-7

IUPAC Name: N,N-diethyl-2-(naphthalen-1-yloxy)propanamide
Synonyms: N,N-diethyl-2-(naphthalen-1-yloxy)propanamide
Molecular Weight (g/mol): 271.36
Molecular Formula: C17H21NO2
InChi Key: WXZVAROIGSFCFJ-UHFFFAOYNA-N
SMILES: CCN(CC)C(=O)C(C)OC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results